Learn More
Description
- Reagent for Coulometric Karl Fischer Titrations.
- Catholyte containing halogenated hydrocarbons for oily samples.
- Free of pyridine. For use with all coulometric titrators.
Specifications
Specifications
| Color | Colorless to Yellow |
| Physical Form | Liquid |
| CAS | 7553-56-2 |
| Quantity | 25 mL |
| Packaging | Amber Glass Bottle |
| UN Number | 1993 |
| Chemical Name or Material | AquaStar™ Coulomat C, Original Formulation |
Safety and Handling
| DOTInformation | Class 3: Flammable Liquid, Toxic |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.