Learn More
Andwin Scientific AQUASTAR LACTOSE STD .5% 10G
SDP
Supplier:  Andwin Scientific EM1.12939.0010
Encompass_Preferred
The Aquastar Lactose Standard has a water content of about 5% and a solubility in methanol that allow this product to be used for coulometry and volumetric Karl Fischer titrations. As an oven standard, it can work under the temperature of 140–160°C. It is sold in a 10 g poly bottle.
Catalog No. NC9691444