Learn More
Cayman Chemical AquastatIn A 2.5mg
SDP
Catalog No. 502950599
Encompass_Preferred
A fungal metabolite; active against S. aureus (MIC = 32 UG/ml); inhibits S. aureus fatty acid synthesis (IC50 = 3.5 µM); inhibits S. aureus Fabl (IC50 = 3.2 µM); inhibits the Na+/K+-ATPase and H+/K+-ATPase (IC50s = 7.1 and 6.2 µM, respectively), as well as PTP1B (IC50 = 0.19 µM)
Catalog No. 50-295-0599 Supplier Cayman Chemical Supplier No. 332532.5 MG
A fungal metabolite; active against S. aureus (MIC = 32 UG/ml); inhibits S. aureus fatty acid synthesis (IC50 = 3.5 µM); inhibits S. aureus Fabl (IC50 = 3.2 µM); inhibits the Na+/K+-ATPase and H+/K+-ATPase (IC50s = 7.1 and 6.2 µM, respectively), as well as PTP1B (IC50 = 0.19 µM)