Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Aquatic Detectives: Solve the Mystery of the Contaminated Waterway
SDPSCI_ED

Catalog No. S05966 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05966 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05966 Supplier EDVOTEK™ Supplier No. 929
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this 'Aquatic Detectives' kit experiment, students will investigate water pollution and test simulated water samples to determine what contamination is present. They will then determine which factories in the narrative contributed the pollutants.

  • Introduce students to the importance of clean water for aquatic and terrestrial ecosystem diversity
  • Perform water quality investigations to determine the site of water pollution
  • Discuss real world problems and identify solutions
  • Group Size: For 10 Lab Groups
  • Time Required: Complete in 45 minutes
  • Kit Includes: Instructions, simulated water pollutants, simulated indicators. microcentrifuge tubes, transfer pipettes, and tube labels
  • All You Need: PPE, distilled water and assorted laboratory glassware. Optional micropipettes with tips
  • Storage: Room Temperature Stable

Specifications

Grade 8 to Undergraduate
Includes Instructions, simulated water pollutants, simulated indicators. microcentrifuge tubes, transfer pipettes, and tube labels
Class Size 10 Lab Groups
Product Type Water Testing Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.