Learn More
AlphaProTech AquaTrak™ Ankle High Boot Cover
Catalog No. 19811664 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Large
Medium
Universal
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-811-664 Large
19-811-661 Universal
19-811-663 Medium
19-811-662 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-811-664 Supplier AlphaProTech Supplier No. BT914B6B
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
AquaTrak™ Ankle High Boot Covers feature superior traction in wet and dry conditions.
  • Superior Anti-Skid Properties
  • Seamless Sole Design
  • Low Particle Count
  • Sonic Welded Seams
Specifications
Size Large
Material Polyethylene, Polypropylene Blend
Color Blue
Closure Type Elastic Top with Ankle Ties
Sole Type Anti-skid