Learn More
Cooper Atkins Corporation AQUATUFF WATERPROOF TC INST
Catalog No. NC9069061
Encompass_Preferred
The AquaTuff 35100-K model is a high accuracy instrument designed to withstand the harsh environments in a kitchen or in food processing. The ABS housing is extremely durable and IPX7 waterproof rated. The housing is ergonomic with a tapered design and curved back to fit nicely in your hand. The 35100-K can be used with any Type K thermocouple probe with a standard mini-connector. Temperatur Range: -100 to 999F (-73 to 537C).
Catalog No. NC9069061 Supplier Cooper Atkins Corporation Supplier No. 35100K
The AquaTuff 35100-K model is a high accuracy instrument designed to withstand the harsh environments in a kitchen or in food processing. The ABS housing is extremely durable and IPX7 waterproof rated. The housing is ergonomic with a tapered design and curved back to fit nicely in your hand. The 35100-K can be used with any Type K thermocouple probe with a standard mini-connector. Temperatur Range: -100 to 999F (-73 to 537C).