Learn More
Innovive Inc Chlorinated Water - Mouse - 300ml
Catalog No. NC1792211
Encompass
This item is sold in quantities of 1 pallet (1848 units) per supplier requirements. 100% PET plastic water bottle pre-filled with chlorinated water. This water has been chlorinated to 1.0-3.0 ppm and has less than 80 ppb of trihalomethanes.
Catalog No. NC1792211 Supplier Innovive Inc Supplier No. MWB300CL
This item is sold in quantities of 1 pallet (1848 units) per supplier requirements. 100% PET plastic water bottle pre-filled with chlorinated water. This water has been chlorinated to 1.0-3.0 ppm and has less than 80 ppb of trihalomethanes.