Learn More
Innovive Inc Ultrapure Water - Mouse - 300ml
Catalog No. NC2158833
Encompass
This item has a minimum order quantity of 1848 per supplier requirements. Plastic mouse water bottle pre-filled with ultra-purified water (has less than 0.1 CFU/ml and less than 500 ppb of total organic carbon).
This item is not returnable. View return policy
Catalog No. NC2158833 Supplier Innovive Inc Supplier No. MWB300PL
This item has a minimum order quantity of 1848 per supplier requirements. Plastic mouse water bottle pre-filled with ultra-purified water (has less than 0.1 CFU/ml and less than 500 ppb of total organic carbon).