Learn More
Innovive Inc Ultrapure Water - Mouse - 300ml
Supplier:  Innovive Inc MWB300PS
Encompass
This item has a minimum order quantity of 924 per supplier requirements. Plastic mouse water bottle pre-filled with ultra-purified water (has less than 0.1 CFU/ml and less than 500 ppb of total organic carbon).
Catalog No. NC2136949