Learn More
Electron Microscopy Sciences CryoJane Aqueous Buffer Salt Mix
SDP
Catalog No. 5030405
Encompass
Used in the preparation of the aqueous fixative, which preserves morphological detail in frozen sections. 2 vials per package, (each vial makes 180 ml of buffer). For use with the CryoJane Tape-Transfer System.
Catalog No. 50-304-05 Supplier Electron Microscopy Sciences Supplier No. 6280076
Used in the preparation of the aqueous fixative, which preserves morphological detail in frozen sections. 2 vials per package, (each vial makes 180 ml of buffer). For use with the CryoJane Tape-Transfer System.