Learn More
Electron Microscopy Sciences Glutaraldehyde 70% Aqueous Solution, 111-30-8, MFCD00007025, 10 mL ampoule
SDP
Catalog No. 5026193
Encompass_Preferred
Formula: OCHCH2CH2CH2CHO, MW: 100.12. After glutaraldehyde has been purified to EM grade, it has a high risk of polymerizing if it is not handled properly. Electron Microscopy Sciences takes special precautions to assure a polymer-free, pure glutaraldehyde. 4 to (-18)°C.
Catalog No. 50-261-93 Supplier Electron Microscopy Sciences Supplier No. 16350
Formula: OCHCH2CH2CH2CHO, MW: 100.12. After glutaraldehyde has been purified to EM grade, it has a high risk of polymerizing if it is not handled properly. Electron Microscopy Sciences takes special precautions to assure a polymer-free, pure glutaraldehyde. 4 to (-18)°C.