Learn More
Biogenex Laboratories AQUEOUS MOUNTING MEDIUM
SDP
Catalog No. NC9674638
Encompass_Preferred
One bottle (15 ml) of Aqueous Mounting Medium, containing gelatin, glycerol and Sodium azide
Catalog No. NC9674638 Supplier Biogenex Laboratories Supplier No. HK0995K
One bottle (15 ml) of Aqueous Mounting Medium, containing gelatin, glycerol and Sodium azide