Learn More
Waters Corp Aqueous Seal Kit (100 µL) 2/pk
Catalog No. 50793523 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Aqueous Seal is intended for buffered/aqueous applications, particularly when there is a high water content. The seal is made of high molecular weight polyethylene and has a PEEK insert to help guide the plunger, reducing the side loads on the sealing material.
Catalog No. 50-793-523 Supplier Waters Corp Supplier No. WAT025296
The Aqueous Seal is intended for buffered/aqueous applications, particularly when there is a high water content. The seal is made of high molecular weight polyethylene and has a PEEK insert to help guide the plunger, reducing the side loads on the sealing material.