Learn More
Waters Corp Waters Aqueous Seal Kit, 100 μL, 4/pk, High Molecular Weight Polyethylene
Catalog No. 50793524 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Aqueous Seal is intended for buffered/aqueous applications, particularly when there is a high water content. The seal is made of high molecular weight polyethylene and has a PEEK insert to help guide the plunger, reducing the side loads on the sealing material.
  • Intended for buffered/aqueous applications
  • Suitable for applications with high water content
  • Made of high molecular weight polyethylene
  • Features a PEEK insert to help guide the plunger
  • Reduces side loads on the sealing material
  • Contributes to improved performance and longevity
Catalog No. 50-793-524 Supplier Waters Corp Supplier No. WAT025297
The Aqueous Seal is intended for buffered/aqueous applications, particularly when there is a high water content. The seal is made of high molecular weight polyethylene and has a PEEK insert to help guide the plunger, reducing the side loads on the sealing material.
  • Intended for buffered/aqueous applications
  • Suitable for applications with high water content
  • Made of high molecular weight polyethylene
  • Features a PEEK insert to help guide the plunger
  • Reduces side loads on the sealing material
  • Contributes to improved performance and longevity