Learn More
Waters Corp Waters Corporation Flowcell, ACQUITY PDA, 10 mm, 250 nL
Catalog No. 50179744 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This flow cell assembly is designed for use with ACQUITY UPLC PDA Detector and ACQUITY UPLC PDA eλ Detector systems.
  • Flow cell dimensions are 10 mm and 250 nL.
  • Compatible with ACQUITY UPLC PDA Detector and ACQUITY UPLC PDA eλ Detector.
  • Includes flow cell assembly.
  • Includes tubing to flow cell (0.0025 ID PEEK NUT PDA).
  • Includes tubing to flow cell (0.004 ID LT PEEK NUT PDA).
Catalog No. 50-179-744 Supplier Waters Corp Supplier No. 205015035
Add to Cart
Add to Cart
Request a Quote
This flow cell assembly is designed for use with ACQUITY UPLC PDA Detector and ACQUITY UPLC PDA eλ Detector systems.
  • Flow cell dimensions are 10 mm and 250 nL.
  • Compatible with ACQUITY UPLC PDA Detector and ACQUITY UPLC PDA eλ Detector.
  • Includes flow cell assembly.
  • Includes tubing to flow cell (0.0025 ID PEEK NUT PDA).
  • Includes tubing to flow cell (0.004 ID LT PEEK NUT PDA).