Learn More
Sigma Aldrich Aquivion™ D98-25BS
Catalog No. 502908096
Encompass
Change view
Click to view available options
Quantity:
250 mL
Catalog No. Quantity
50-290-8096 250 mL
1 options
Catalog No. 50-290-8096 Supplier Sigma Aldrich Supplier No. 802557250ML
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Aquivion(R) PFSA dispersions are based on the short-side-chain (SSC) copolymer of Tetrafluoroethylene and the Sulfonyl Fluoride Vinyl Ether (SFVE) CF2=CF2-O-(CF2)2-SO2F industrially produced by Solvay Specialty Polymers. The resulting perfluoropolymer's functional groups are in their sulfonic acid form, SO3H.Can be used as a formulation additive for production of polymer electrolyte fuel cell electrodes, manufacture or repair of cation-exchange membranes, surface coatings of PTFE membranes to improve their wetting behavior or for the development of superacid catalysts. Addition of further film-forming and/or other volatile or non-volatile chemicals is necessary to impart the desired processing and end-product characteristics.Aquivion is a registered trademark of Solvay Group
TRUSTED_SUSTAINABILITY
Specifications
Synonym Aquivion™ SO3H-form; Tetrafluoroethylene-perfluoro(3-oxa-4-pentenesulfonic acid) copolymer, Ethanesulfonic acid
Quantity 250 mL