Learn More
Selleck Chemical LLC AQX-016A S3486-5mg
SDP
Catalog No. 503108330
Encompass_Preferred
AQX-016A is a potent agonist of SHIP1 AQX-016A inhibits DNFB induced inflammation in a mouse ear edema/cutaneous anaphylaxis model
Catalog No. 50-310-8330 Supplier Selleck Chemical LLC Supplier No. S34865MG
AQX-016A is a potent agonist of SHIP1 AQX-016A inhibits DNFB induced inflammation in a mouse ear edema/cutaneous anaphylaxis model