Learn More
Medchemexpress LLC AR-13324 analog (mesylate) | 2095432-73-6 | 98.0% | 631.72 | 10 MM * 1 ML
SDP
Catalog No. 5000384915
Encompass_Preferred
AR-13324 analog mesylate is an analog of AR-13324. AR-13324 is an ROCK and norepinephrine transporter (NET) inhibitor.
  • 10 mM * 1 mL in DMSO (ready for reconstitution)
  • 10 mM * 1 mL in DMSO
  • 5 mg
  • 10 mg
  • 50 mg
  • 100 mg
  • 200 mg
Catalog No. 50-003-84915 Supplier Medchemexpress LLC Supplier No. HY1934610MM/1ML
Add to Cart
Add to Cart
AR-13324 analog mesylate is an analog of AR-13324. AR-13324 is an ROCK and norepinephrine transporter (NET) inhibitor.
  • 10 mM * 1 mL in DMSO (ready for reconstitution)
  • 10 mM * 1 mL in DMSO
  • 5 mg
  • 10 mg
  • 50 mg
  • 100 mg
  • 200 mg