Learn More
Medchemexpress LLC AR-13324 analog mesylate | 2095432-73-6 | 98.0% | 631.72 | 50 MG
SDP
Catalog No. 5000385058
Encompass_Preferred
AR-13324 analog mesylate is an analog of AR-13324. AR-13324 is an ROCK and norepinephrine transporter (NET) inhibitor.
  • Analog of AR-13324.
  • Ar-13324 is a rock and norepinephrine transporter (net) inhibitor.
  • Available in various sizes, including 50 mg.
Catalog No. 50-003-85058 Supplier Medchemexpress LLC Supplier No. HY1934650MG
AR-13324 analog mesylate is an analog of AR-13324. AR-13324 is an ROCK and norepinephrine transporter (NET) inhibitor.
  • Analog of AR-13324.
  • Ar-13324 is a rock and norepinephrine transporter (net) inhibitor.
  • Available in various sizes, including 50 mg.