Learn More
Medchemexpress LLC AR-13324 analog mesylate | 2095432-73-6 | 98.0% | 631.72 | C29H33N3O9S2 | 10 MG
SDP
Catalog No. 5000294756
Encompass_Preferred
AR-13324 analog mesylate is a research-grade mesylate salt analog of AR-13324, a Rho-associated kinase (ROCK) and norepinephrine transporter (NET) inhibitor. It is supplied as a light brown to brown liquid intended for laboratory research and assay development.
  • Research-use compound related to ROCK and NET inhibition.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Molecular weight 631.72 g/mol for formulation calculations.
  • Supplied as a mesylate salt in a liquid form for easy handling.
  • Store sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-94756 Supplier Medchemexpress LLC Supplier No. HY1934610MG
AR-13324 analog mesylate is a research-grade mesylate salt analog of AR-13324, a Rho-associated kinase (ROCK) and norepinephrine transporter (NET) inhibitor. It is supplied as a light brown to brown liquid intended for laboratory research and assay development.
  • Research-use compound related to ROCK and NET inhibition.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Molecular weight 631.72 g/mol for formulation calculations.
  • Supplied as a mesylate salt in a liquid form for easy handling.
  • Store sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.