Learn More
Abcam AR 231453, GPR119 agonist, 25MG
Catalog No. 502706494
Encompass_Preferred
MW 505.5 Da, Purity >99%. Potent, selective G-protein coupled receptor 119 (GPR119) agonist (EC₅₀ = 0.68 nM). Enhances glucose-induced insulin release. Antidiabetic activity. Orally active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6494 Supplier Abcam Supplier No. AB14162725MG
MW 505.5 Da, Purity >99%. Potent, selective G-protein coupled receptor 119 (GPR119) agonist (EC₅₀ = 0.68 nM). Enhances glucose-induced insulin release. Antidiabetic activity. Orally active in vivo.
The product is subject to the following: Abcam Restricted Use Statement