Learn More
Apexbio Technology LLC AR-42 (OSU-HDAC42) 935881-37-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061445
Encompass_Preferred
AR-42 (OSU-HDAC42 CAS 935881-37-1) is a hydroxamate-derived small molecule structurally based on phenylbutyrate functioning as a potent histone deacetylase (HDAC) inhibitor It modulates chromatin structure by inhibiting HDAC activity (IC50 16 nM) thus increasing histone H3 acetylation -tubulin acetylation and expression of p21 markers indicative of HDAC inhibition AR-42 also affects apoptotic and survival pathway regulators including Akt Bcl-xL Bax Ku70 and Survivin and likely acts partially through suppression of the PI3K/Akt signaling axis It shows antitumor activity in prostate liver and various other cancer models supporting its utility for cancer and epigenetics research
Catalog No. 50-000-61445 Supplier Apexbio Technology LLC Supplier No. A41045.1
AR-42 (OSU-HDAC42 CAS 935881-37-1) is a hydroxamate-derived small molecule structurally based on phenylbutyrate functioning as a potent histone deacetylase (HDAC) inhibitor It modulates chromatin structure by inhibiting HDAC activity (IC50 16 nM) thus increasing histone H3 acetylation -tubulin acetylation and expression of p21 markers indicative of HDAC inhibition AR-42 also affects apoptotic and survival pathway regulators including Akt Bcl-xL Bax Ku70 and Survivin and likely acts partially through suppression of the PI3K/Akt signaling axis It shows antitumor activity in prostate liver and various other cancer models supporting its utility for cancer and epigenetics research