Learn More
Apexbio Technology LLC AR-42 (OSU-HDAC42), 2mg. Cas: 935881-37-1 MFCD: MFCD17676151. HDAC inhibitor,novel and potent
SDP
Supplier:  Apexbio Technology LLC A41042
Encompass_Preferred
AR-42 (also known as OSU-HDAC42), a derivative of hydroxamate-tethered phenylbutyrate, is a novel and potent inhibitor of histone deacetylase (HDAC) that potently inhibits the activity of HDAC with 50% inhibition concentration IC50 value of 16 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4881