Learn More
Santa Cruz Biotechnology AR(441) ANTIBODY
Supplier:  Santa Cruz Biotechnology SC7305
Encompass
AR (441),mouse monoclonal IgG1; 200 microgram/ml,raised against amino acids 299-315 of androgen receptor of human origin,recommended for detection of androgen receptor of human origin by WB, IP and IHC(P),ImmunoCruz Staining System
Catalog No. NC9745836