Learn More
TARGETMOL CHEMICALS INC AR-A014418 50MG
SDP
Catalog No. 502581067
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AR-A014418 (GSK 3(beta) inhibitor VIII) is an ATP-competitive and selective GSK3(beta) inhibitor. purity: 99%
Catalog No. 50-258-1067 Supplier TARGETMOL CHEMICALS INC Supplier No. T188150MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AR-A014418 (GSK 3(beta) inhibitor VIII) is an ATP-competitive and selective GSK3(beta) inhibitor. purity: 99%