Learn More
Abcam AR-A014418, GSK-3 inhibitor, 50MG
Catalog No. 502706707
Encompass_Preferred
MW 308.32 Da, Purity >99%. Highly selective GSK-3 inhibitor (IC₅₀ = 104 nM). ATP competitive (Ki = 38 nM). Selective over cdc2, cdc5 and 26 other kinases. Inhibits tau phosphorylation (ser 396) at a GSK-3 specific site. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6707 Supplier Abcam Supplier No. AB14129550MG
MW 308.32 Da, Purity >99%. Highly selective GSK-3 inhibitor (IC₅₀ = 104 nM). ATP competitive (Ki = 38 nM). Selective over cdc2, cdc5 and 26 other kinases. Inhibits tau phosphorylation (ser 396) at a GSK-3 specific site. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement