Learn More
INTACT GENOMICS INC Ar.A4 Agrobacterium ElectroCompetent Cells 6x50µl
Supplier:  INTACT GENOMICS INC 127212
Encompass
Intact Genomics (ig) Ar.A4 Agrobacterium Electrocompetent Cells are made from a specific strain of Rhizobium rhizogenes (formerly Agrobacterium rhizogenes), Agrobacterium rhizogenes (kan R) Ar A4 Ri (agropine type). Agrobacterium rhizogenes is a soil-borne gram-negative bacterium that can infect most dicotyledons, a few monocotyledons, and some gymnosperms. Ar.A4 Electrocompetent Agrobacterium are optimized for the highest transformation efficiencies and are useful for transgenic operations of corn, tobacco, carrot, and other plants. Ar.A4 Agrobacterium rhizogenes strain contains agrobacterium-type Ri plasmid and displays kanamycin resistance.Product Includes and Storage:Ar. A4 Agrobacterium Electrocompetent Cells: -80 ºCpIG7-spe control DNA: -20 ºCAgro Recovery medium: 4 ºC
Catalog No. 50-000-83040