Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 367MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: SPM335. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: LNCap cells or Prostate carcinoma. Cellular Staining: Nuclear. Buffer: 1mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide, aa 299-315, (STEDTAEYSPFKGGYTK) of human AR. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 110kDa.Chromosome: Xq12. Gene Symbol: AR. Gene ID: 367. Ensemble: ENSG00000169083. Uniprot: NP_001334990.
Catalog No. 50-188-4836