Learn More
Aviva Systems Biology Corp AR Antibody - N-terminal region (ARP30974_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30974_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human AR. Alternate names: AIS, DHTR, HUMARA, KD, NR3C4, SBMA, SMAX1, TFM, HYSP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54174