Learn More
Medchemexpress LLC AR-C102222 hydrochloride | 1781934-50-6 | 99.8% | 10 MG
SDP
Catalog No. 5000233882
Encompass_Preferred
AR-C102222 hydrochloride is the hydrochloride salt of AR-C102222, a potent, competitive, orally active, and highly selective inducible nitric oxide synthase (iNOS) inhibitor (IC50 = 37 nM). It is supplied as a white to off-white solid for research use.
  • Potent, selective iNOS inhibitor (IC50 = 37 nM).
  • White to off-white solid with high purity.
  • Soluble in DMSO; may require sonication and warming.
  • Store sealed at 4°C; solutions stable at -20°C to -80°C for extended storage.
  • Available in small research pack sizes suitable for laboratory use.
Catalog No. 50-002-33882 Supplier Medchemexpress LLC Supplier No. HY12122A10MG
AR-C102222 hydrochloride is the hydrochloride salt of AR-C102222, a potent, competitive, orally active, and highly selective inducible nitric oxide synthase (iNOS) inhibitor (IC50 = 37 nM). It is supplied as a white to off-white solid for research use.
  • Potent, selective iNOS inhibitor (IC50 = 37 nM).
  • White to off-white solid with high purity.
  • Soluble in DMSO; may require sonication and warming.
  • Store sealed at 4°C; solutions stable at -20°C to -80°C for extended storage.
  • Available in small research pack sizes suitable for laboratory use.