Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AR-C102222 hydrochloride | 1781934-50-6 | 99.8% | 50 MG
SDP

Catalog No. 5000231742
Encompass_Preferred

AR-C102222 hydrochloride is a selective inducible nitric oxide synthase (iNOS) inhibitor (IC50 = 37 nM) provided as the hydrochloride salt for research use. It exhibits reported antinociceptive and anti-inflammatory activity and is supplied as a high-purity solid suitable for in vitro and in vivo studies.

  • Purity 99.83%.
  • Molecular weight 418.83 g/mol.
  • Chemical formula C19H17ClF2N6O.
  • Soluble in DMSO (100 mg/mL); formulatable for in vivo dosing (≥5 mg/mL in recommended vehicles).
  • Storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available as solid in multiple sizes, including 50 mg.

Catalog No. 50-002-31742 Supplier Medchemexpress LLC Supplier No. HY12122A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AR-C102222 hydrochloride is a selective inducible nitric oxide synthase (iNOS) inhibitor (IC50 = 37 nM) provided as the hydrochloride salt for research use. It exhibits reported antinociceptive and anti-inflammatory activity and is supplied as a high-purity solid suitable for in vitro and in vivo studies.

  • Purity 99.83%.
  • Molecular weight 418.83 g/mol.
  • Chemical formula C19H17ClF2N6O.
  • Soluble in DMSO (100 mg/mL); formulatable for in vivo dosing (≥5 mg/mL in recommended vehicles).
  • Storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available as solid in multiple sizes, including 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.