Learn More
Selleck Chemical LLC AR-C155858-1mg
SDP
Catalog No. 502389868
Encompass_Preferred
AR-C155858 is a selective monocarboxylate transporter MCT1 and MCT2 inhibitor with Ki value of 2.3 nM and 10 nM, respectively.
Catalog No. 50-238-9868 Supplier Selleck Chemical LLC Supplier No. S79191MG
AR-C155858 is a selective monocarboxylate transporter MCT1 and MCT2 inhibitor with Ki value of 2.3 nM and 10 nM, respectively.