Learn More
Medchemexpress LLC Ar-m 1000390 (hydrochloride) | 209808-47-9 | MFCD20926357 | 98.2% | 384.94 g/mol | C23H29ClN2O | 10 MG
SDP
Catalog No. 5000212025
Encompass_Preferred
AR-M 1000390 hydrochloride is a selective and potent δ-opioid receptor agonist supplied as the hydrochloride salt for research use. It exhibits an EC50 of 7.2 ± 0.9 nM for δ agonist potency, a reported purity of 98.2%, and a molecular weight of 384.94 g/mol. The compound is provided as a white to off-white solid intended for in vitro and in vivo pharmacology studies.
  • Selective δ-opioid receptor agonist for receptor pharmacology studies.
  • Potent activity with EC50 7.2 ± 0.9 nM for δ agonist potency.
  • High reported purity (98.2%), suitable for research assays.
  • Solid form, white to off-white, convenient for handling and formulation.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes, including 10 mg.
Catalog No. 50-002-12025 Supplier Medchemexpress LLC Supplier No. HY101039A10MG
AR-M 1000390 hydrochloride is a selective and potent δ-opioid receptor agonist supplied as the hydrochloride salt for research use. It exhibits an EC50 of 7.2 ± 0.9 nM for δ agonist potency, a reported purity of 98.2%, and a molecular weight of 384.94 g/mol. The compound is provided as a white to off-white solid intended for in vitro and in vivo pharmacology studies.
  • Selective δ-opioid receptor agonist for receptor pharmacology studies.
  • Potent activity with EC50 7.2 ± 0.9 nM for δ agonist potency.
  • High reported purity (98.2%), suitable for research assays.
  • Solid form, white to off-white, convenient for handling and formulation.
  • Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes, including 10 mg.