Learn More
Medchemexpress LLC AR-M 1000390 hydrochloride | 209808-47-9 | MFCD20926357 | 98.2% | 384.94 g/mol | C23H29ClN2O | 50 MG
SDP
Catalog No. 5000213592
Encompass_Preferred
AR-M 1000390 hydrochloride is a potent, highly selective small-molecule agonist of the δ (delta) opioid receptor provided as the hydrochloride salt for research use. It exhibits low-nanomolar potency, is supplied as a white to off-white solid, and is commonly used in in vitro receptor pharmacology and selectivity profiling.
  • Potent δ-opioid receptor agonist with EC50 of 7.2 ± 0.9 nM.
  • High purity suitable for pharmacological studies (~98.2%).
  • Provided as a stable hydrochloride solid, white to off-white.
  • Available in multiple milligram sizes for assay flexibility.
  • Suitable for in vitro receptor pharmacology and selectivity profiling.
  • Recommended sealed storage away from moisture; stable refrigerated.
Catalog No. 50-002-13592 Supplier Medchemexpress LLC Supplier No. HY101039A50MG
AR-M 1000390 hydrochloride is a potent, highly selective small-molecule agonist of the δ (delta) opioid receptor provided as the hydrochloride salt for research use. It exhibits low-nanomolar potency, is supplied as a white to off-white solid, and is commonly used in in vitro receptor pharmacology and selectivity profiling.
  • Potent δ-opioid receptor agonist with EC50 of 7.2 ± 0.9 nM.
  • High purity suitable for pharmacological studies (~98.2%).
  • Provided as a stable hydrochloride solid, white to off-white.
  • Available in multiple milligram sizes for assay flexibility.
  • Suitable for in vitro receptor pharmacology and selectivity profiling.
  • Recommended sealed storage away from moisture; stable refrigerated.