Learn More
Medchemexpress LLC AR-R17779 hydrochloride | 178419-42-6 | 99.0% | C9H15ClN2O2 | 10MG
SDP
Catalog No. 5000259704
Encompass_Preferred
AR-R17779 hydrochloride is a research-grade, potent and selective full agonist of the α7 subtype of neuronal nicotinic acetylcholine receptors, provided as the hydrochloride salt for preclinical pharmacology studies. It is intended for in vitro and in vivo research to probe receptor signaling, cognitive effects, and neuroprotective mechanisms.
  • Potent, selective α7 nicotinic acetylcholine receptor agonist.
  • High purity (99.0%) suitable for research applications.
  • Supplied as a white to off-white solid for ease of handling.
  • Stable under recommended storage conditions to preserve activity.
  • Suitable for in vitro assays and in vivo pharmacology studies.
Catalog No. 50-002-59704 Supplier Medchemexpress LLC Supplier No. HY135483A10MG
AR-R17779 hydrochloride is a research-grade, potent and selective full agonist of the α7 subtype of neuronal nicotinic acetylcholine receptors, provided as the hydrochloride salt for preclinical pharmacology studies. It is intended for in vitro and in vivo research to probe receptor signaling, cognitive effects, and neuroprotective mechanisms.
  • Potent, selective α7 nicotinic acetylcholine receptor agonist.
  • High purity (99.0%) suitable for research applications.
  • Supplied as a white to off-white solid for ease of handling.
  • Stable under recommended storage conditions to preserve activity.
  • Suitable for in vitro assays and in vivo pharmacology studies.