Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AR-R17779 hydrochloride | 178419-42-6 | MFCD20926337 | 99.0% | 218.68 | C9H15ClN2O2 | 5 MG
SDP

Catalog No. 5000223503
Encompass_Preferred

AR-R17779 hydrochloride is a selective agonist of α7 nicotinic acetylcholine receptors supplied as the hydrochloride salt for laboratory research. It has been reported to influence learning and memory, show anxiolytic activity, and modulate anti-inflammatory cholinergic pathways. Purity is typically 99.0%.

  • Selective α7 nicotinic receptor agonist.
  • Provided as the hydrochloride salt.
  • Used in neuroscience and pharmacology research.
  • Reported effects on learning, memory, anxiety, and inflammation pathways.
  • High purity (99.0%).
  • Molecular weight 218.68; formula C9H15ClN2O2.
  • Available in small pack sizes for laboratory use.

Catalog No. 50-002-23503 Supplier Medchemexpress LLC Supplier No. HY135483A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AR-R17779 hydrochloride is a selective agonist of α7 nicotinic acetylcholine receptors supplied as the hydrochloride salt for laboratory research. It has been reported to influence learning and memory, show anxiolytic activity, and modulate anti-inflammatory cholinergic pathways. Purity is typically 99.0%.

  • Selective α7 nicotinic receptor agonist.
  • Provided as the hydrochloride salt.
  • Used in neuroscience and pharmacology research.
  • Reported effects on learning, memory, anxiety, and inflammation pathways.
  • High purity (99.0%).
  • Molecular weight 218.68; formula C9H15ClN2O2.
  • Available in small pack sizes for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.