Learn More
MSC Annovi Reverberi Triplex Plunger Pump, 2600 PSI, 2.5 GPM
Catalog No. 502815874
Encompass
This D version 3/4 inch hollow shaft triplex pump is designed for direct or pulley driving. It features a brass pump head suitable for a variety of conditions, delivering up to 2600 PSI and 2.5 GPM at 3400 RPM. Constructed with frictionless aluminum connecting rods and ceramic pistons, it offers a compact design.
  • Reliable, high-performance pump designed for consistent and powerful output
  • Brass pump head suitable for a wide range of conditions
  • Frictionless aluminum connecting rods and ceramic pistons for durability and performance
  • Sleek compact design, weighing 15 lbs
  • Direct or pulley driving for versatile installation options
Catalog No. 50-281-5874 Supplier MSC Supplier No. 03013752
This D version 3/4 inch hollow shaft triplex pump is designed for direct or pulley driving. It features a brass pump head suitable for a variety of conditions, delivering up to 2600 PSI and 2.5 GPM at 3400 RPM. Constructed with frictionless aluminum connecting rods and ceramic pistons, it offers a compact design.
  • Reliable, high-performance pump designed for consistent and powerful output
  • Brass pump head suitable for a wide range of conditions
  • Frictionless aluminum connecting rods and ceramic pistons for durability and performance
  • Sleek compact design, weighing 15 lbs
  • Direct or pulley driving for versatile installation options