Learn More
Cayman Chemical -ar-Turmeron 10mg
SDP
Catalog No. 502953704
Encompass_Preferred
An aromatic compound that stimulates the proliferation of peripheral blood mononuclear cells and increases the production of TNF-α, IL-2, and IFN-γ; promotes the maturation of dendritic cells and induces the proliferation of neural stem cells both in vitro and in vivo
Catalog No. 50-295-3704 Supplier Cayman Chemical Supplier No. 1175910 MG
An aromatic compound that stimulates the proliferation of peripheral blood mononuclear cells and increases the production of TNF-α, IL-2, and IFN-γ; promotes the maturation of dendritic cells and induces the proliferation of neural stem cells both in vitro and in vivo