Learn More
Medchemexpress LLC ar-Turmerone | 532-65-0 | 5 MG
SDP
Catalog No. 5000418370
Encompass_Preferred
ar-Turmerone is an orally active and major bioactive compound derived from the herb Curcuma longa. It exhibits anti-tumorigenesis and anti-inflammatory activities, making it suitable for various research applications, particularly in studies related to neurological disorders.
  • Orally active compound
  • Major bioactive compound of Curcuma longa
  • Exhibits anti-tumorigenesis activity
  • Exhibits anti-inflammatory activity
  • Induces apoptosis in U937 cells
  • Positively modulates murine DCs
  • Induces NSC proliferation in vitro and in vivo
  • Suitable for neurologic disorders study
Catalog No. 50-004-18370 Supplier Medchemexpress LLC Supplier No. HYN67035MG
ar-Turmerone is an orally active and major bioactive compound derived from the herb Curcuma longa. It exhibits anti-tumorigenesis and anti-inflammatory activities, making it suitable for various research applications, particularly in studies related to neurological disorders.
  • Orally active compound
  • Major bioactive compound of Curcuma longa
  • Exhibits anti-tumorigenesis activity
  • Exhibits anti-inflammatory activity
  • Induces apoptosis in U937 cells
  • Positively modulates murine DCs
  • Induces NSC proliferation in vitro and in vivo
  • Suitable for neurologic disorders study