Learn More
TARGETMOL CHEMICALS INC AR42 10MG
SDP
Catalog No. 502579392
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AR42 (OSU-HDAC42) is an HDAC inhibitor (IC50 30 nM). purity: 99%
Catalog No. 50-257-9392 Supplier TARGETMOL CHEMICALS INC Supplier No. T639210MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AR42 (OSU-HDAC42) is an HDAC inhibitor (IC50 30 nM). purity: 99%