Learn More
Medchemexpress LLC AR420626 100mg | 1798310-55-0 | 417.29 g·mol^-1 | C21H18Cl2N2O3 | 100 MG
SDP
Catalog No. 5000227479
Encompass_Preferred
AR420626 is a selective agonist of free fatty acid receptor 3 (FFAR3/GPR41) used in receptor pharmacology and signaling research. It is supplied as an off-white to light yellow solid with reported purity ≥98% and a molecular weight of 417.29 g·mol⁻¹. Recommended storage and handling conditions depend on form (powder or solution) to preserve stability.
Catalog No. 50-002-27479 Supplier Medchemexpress LLC Supplier No. HY116522100MG
AR420626 is a selective agonist of free fatty acid receptor 3 (FFAR3/GPR41) used in receptor pharmacology and signaling research. It is supplied as an off-white to light yellow solid with reported purity ≥98% and a molecular weight of 417.29 g·mol⁻¹. Recommended storage and handling conditions depend on form (powder or solution) to preserve stability.