Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AR420626 10mg | 1798310-55-0 | 417.29 g/mol | C21H18Cl2N2O3 | 10 MG
SDP

Catalog No. 5000227580
Encompass_Preferred

AR-420626 is a selective agonist of free fatty acid receptor 3 (FFAR3/GPR41) used as a research tool to probe FFAR3-mediated signaling. It exhibits an IC50 of 117 nM, is supplied as a solid 10 mg quantity with reported purity of 98.29%, and is intended for research use only.

  • Selective FFAR3 agonist with an IC50 of 117 nM.
  • Reported purity 98.29%.
  • Molecular formula C21H18Cl2N2O3; molecular weight 417.29 g/mol.
  • Supplied as a 10 MG solid suitable for in vitro studies.
  • Recommended storage: powder -20°C (up to 3 years).
  • For research use only; not for human or diagnostic use.

Catalog No. 50-002-27580 Supplier Medchemexpress LLC Supplier No. HY11652210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AR-420626 is a selective agonist of free fatty acid receptor 3 (FFAR3/GPR41) used as a research tool to probe FFAR3-mediated signaling. It exhibits an IC50 of 117 nM, is supplied as a solid 10 mg quantity with reported purity of 98.29%, and is intended for research use only.

  • Selective FFAR3 agonist with an IC50 of 117 nM.
  • Reported purity 98.29%.
  • Molecular formula C21H18Cl2N2O3; molecular weight 417.29 g/mol.
  • Supplied as a 10 MG solid suitable for in vitro studies.
  • Recommended storage: powder -20°C (up to 3 years).
  • For research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.