Learn More
Medchemexpress LLC AR420626 25mg | 1798310-55-0 | 417.29 | C21H18Cl2N2O3 | 25 MG
SDP
Catalog No. 5000228430
Encompass_Preferred
AR420626 is a research compound that acts as a selective agonist of free fatty acid receptor 3 (FFAR3/FFA3). It is used in vitro and in vivo to investigate FFAR3-mediated signaling, inflammatory responses, and metabolic or cancer-related pathways. Reported activity values support its use in pharmacology and cell-based assays.
  • Selective FFAR3 agonist with reported IC50 ≈117 nM.
  • High purity suitable for biological assays and pharmacology studies.
  • Available as a solid or as a DMSO solution for convenient dosing.
  • Multiple package sizes support screening and follow-up studies.
  • Compatible with in vitro and in vivo experimental workflows.
Catalog No. 50-002-28430 Supplier Medchemexpress LLC Supplier No. HY11652225MG
AR420626 is a research compound that acts as a selective agonist of free fatty acid receptor 3 (FFAR3/FFA3). It is used in vitro and in vivo to investigate FFAR3-mediated signaling, inflammatory responses, and metabolic or cancer-related pathways. Reported activity values support its use in pharmacology and cell-based assays.
  • Selective FFAR3 agonist with reported IC50 ≈117 nM.
  • High purity suitable for biological assays and pharmacology studies.
  • Available as a solid or as a DMSO solution for convenient dosing.
  • Multiple package sizes support screening and follow-up studies.
  • Compatible with in vitro and in vivo experimental workflows.