Learn More
Medchemexpress LLC HY-101106 5mg Medchemexpress, AR7 CAS:80306-38-3 Purity:>98%
SDP
Catalog No. 501932041
Encompass_Preferred
Medchemexpress, HY-101106 5mg AR7 CAS:80306-38-3 AR7 is a retinoic acid receptor α ( RARα ) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2041 Supplier Medchemexpress LLC Supplier No. HY1011065MG
Medchemexpress, HY-101106 5mg AR7 CAS:80306-38-3 AR7 is a retinoic acid receptor α ( RARα ) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.