Learn More
Quanterix Inc AR9 Buffer, 10X, 4 X 250 ML
SDP
Catalog No. NC1628008
Encompass
10X buffer for use in Opal™ immunohistochemistry multiplexing protocols. This buffer was developed for use in Opal multicolor immunohistochemistry protocol in microwave treatment (MWT) steps. MWT provides antigen retrieval for formalin-fixed, paraffin embedded (FFPE) tissue, quenches endogenous peroxidase, and removes antibodies after epitopes have been labeled with Opal dyes.
Catalog No. NC1628008 Supplier Quanterix Inc Supplier No. AR9001KT
10X buffer for use in Opal™ immunohistochemistry multiplexing protocols. This buffer was developed for use in Opal multicolor immunohistochemistry protocol in microwave treatment (MWT) steps. MWT provides antigen retrieval for formalin-fixed, paraffin embedded (FFPE) tissue, quenches endogenous peroxidase, and removes antibodies after epitopes have been labeled with Opal dyes.