Learn More
Cayman Chemical ARA 290 (acetate), Purity: ≥95%, 10 mg
SDP
Catalog No. 502910496
Encompass_Preferred
A derivative of EPO; reduces plasma creatine, urea, and AST levels in a rodent model of renal ischemia-reperfusion injury at 0.8 and 8 nmol/kg; i.p.; decreases wound area in a rat model of punch biopsy-induced dermal injury and a rat model of decubitus ulcer; reduces the latency to find the platform in the Morris water maze in a rat model of cortical impact-induced TBI at 30 µg/kg; i.p.; reduces tactile allodynia in a rat model of SNI-induced neuropathic pain
Catalog No. 50-291-0496 Supplier Cayman Chemical Supplier No. 3494310 MG
A derivative of EPO; reduces plasma creatine, urea, and AST levels in a rodent model of renal ischemia-reperfusion injury at 0.8 and 8 nmol/kg; i.p.; decreases wound area in a rat model of punch biopsy-induced dermal injury and a rat model of decubitus ulcer; reduces the latency to find the platform in the Morris water maze in a rat model of cortical impact-induced TBI at 30 µg/kg; i.p.; reduces tactile allodynia in a rat model of SNI-induced neuropathic pain