Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ara h 1 protein, Arachis hypogaea | >90.0% | 100UG
SDP

Catalog No. 5000174155
Encompass_Preferred

Recombinant Ara h 1 protein (mature form K26-N614) from Arachis hypogaea, expressed in E. coli and supplied lyophilized with an N-terminal 6xHis tag for research use. The material is characterized by reducing SDS-PAGE, with an apparent molecular weight of approximately 72 kDa and high purity suitable for immunological and biochemical studies.

  • Recombinant full-length mature Ara h 1 (K26-N614).
  • Expressed in E. coli with an N-terminal 6xHis tag.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Apparent molecular weight approximately 72 kDa by SDS-PAGE.
  • Lyophilized from 0.22 μm filtered PBS with 6% trehalose, pH 7.4.
  • Do not reconstitute below 100 μg/mL in ddH2O.
  • Store at -20 °C; after reconstitution stable at 4 °C for 1 week or -20 °C with carrier; aliquot for long-term storage.
  • Intended for research use only.

Catalog No. 50-001-74155 Supplier Medchemexpress LLC Supplier No. HYP704203100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Ara h 1 protein (mature form K26-N614) from Arachis hypogaea, expressed in E. coli and supplied lyophilized with an N-terminal 6xHis tag for research use. The material is characterized by reducing SDS-PAGE, with an apparent molecular weight of approximately 72 kDa and high purity suitable for immunological and biochemical studies.

  • Recombinant full-length mature Ara h 1 (K26-N614).
  • Expressed in E. coli with an N-terminal 6xHis tag.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Apparent molecular weight approximately 72 kDa by SDS-PAGE.
  • Lyophilized from 0.22 μm filtered PBS with 6% trehalose, pH 7.4.
  • Do not reconstitute below 100 μg/mL in ddH2O.
  • Store at -20 °C; after reconstitution stable at 4 °C for 1 week or -20 °C with carrier; aliquot for long-term storage.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.