Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Allergen Ara h 1 (Arachis hypogaea) | >90.0% | 50UG
SDP

Catalog No. 5000174175
Encompass_Preferred

Ara h 1 is a recombinant full-length mature peanut allergen (Arachis hypogaea) produced in Escherichia coli with an N-terminal 6xHis affinity tag. Supplied as a lyophilized powder and characterized by >90.0% purity, it is suitable for allergen research, antibody development, and assay controls.

  • Full-length mature allergen for immunological assays
  • Expressed in Escherichia coli with N-terminal 6xHis tag for affinity purification
  • Lyophilized powder formulation for convenient storage and transport
  • Purity >90.0% as determined by reducing SDS-PAGE
  • Predicted molecular mass 71.7 kDa
  • Low endotoxin level <1 EU/μg (LAL method)
  • Accession P43237 and gene ID 112711772 provided for reference

Catalog No. 50-001-74175 Supplier Medchemexpress LLC Supplier No. HYP70420350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ara h 1 is a recombinant full-length mature peanut allergen (Arachis hypogaea) produced in Escherichia coli with an N-terminal 6xHis affinity tag. Supplied as a lyophilized powder and characterized by >90.0% purity, it is suitable for allergen research, antibody development, and assay controls.

  • Full-length mature allergen for immunological assays
  • Expressed in Escherichia coli with N-terminal 6xHis tag for affinity purification
  • Lyophilized powder formulation for convenient storage and transport
  • Purity >90.0% as determined by reducing SDS-PAGE
  • Predicted molecular mass 71.7 kDa
  • Low endotoxin level <1 EU/μg (LAL method)
  • Accession P43237 and gene ID 112711772 provided for reference

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.