Learn More
Percival Scientific Arabidopsis Growth Chamber, AR36L3H14, 29.7 cu. ft., SciWhite with red LEDs, 3 tiers, 7-44° C ±0.5°, IntellusUltra controller, touchscreen, standard additive ultrasonic humidifier
SDP
Catalog No. NC3794488
Encompass_Preferred
The AR36L3H14 growth chamber is designed for growth of Arabidopsis and low light plant growth applications. SciWhite with red LEDs are dimmable from 10-100% output with up to 350 µmoles/m2/s irradiance @ 6". Ultrasonic humidifier and standard humidity sensor provide additive humidity. Constructed of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Special airflow insulates shelf level experiments from heat generated by underlying light fixtures. Three tiers of epoxy coated steel wire shelving slide easily on stainless steel rail assemblies. Self-contained air-cooled condensing unit with continuous compressor operation extends life, ensuring precise temperature control by cycling refrigerant and hot gas to coil; prolonging life of compressor, eliminating risk of ice build up in coil. Top mounted refrigeration system allows easy access for servicing. Contact sales@percival-scientific.com for more options.
Catalog No. NC3794488 Supplier Percival Scientific Supplier No. AR36L3H14
Add to Cart
Add to Cart
Request a Quote
The AR36L3H14 growth chamber is designed for growth of Arabidopsis and low light plant growth applications. SciWhite with red LEDs are dimmable from 10-100% output with up to 350 µmoles/m2/s irradiance @ 6". Ultrasonic humidifier and standard humidity sensor provide additive humidity. Constructed of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Special airflow insulates shelf level experiments from heat generated by underlying light fixtures. Three tiers of epoxy coated steel wire shelving slide easily on stainless steel rail assemblies. Self-contained air-cooled condensing unit with continuous compressor operation extends life, ensuring precise temperature control by cycling refrigerant and hot gas to coil; prolonging life of compressor, eliminating risk of ice build up in coil. Top mounted refrigeration system allows easy access for servicing. Contact sales@percival-scientific.com for more options.