Learn More
Percival Scientific Arabidopsis Growth Chamber, AR41L2H11, 37.2 cu. ft., SciWhite with red LEDs, 2 tiers, 7-44° C ±0.5°, IntellusUltra controller, touchscreen, advanced additive ultrasonic humidifier
SDP
Catalog No. NC3794499
Encompass_Preferred
The AR41L2H11 growth chamber is designed for growth of Arabidopsis thaliana and low light plant growth applications. SciWhite with red LEDs are dimmable from 10-100% output with up to 405 µmoles/m2/s irradiance @ 6". Ultrasonic humidifier and advanced humidity sensor provide additive humidity. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Adjustable air diffuser for temperature uniformity. Two white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.
Catalog No. NC3794499 Supplier Percival Scientific Supplier No. AR41L2H11
Add to Cart
Add to Cart
Request a Quote
The AR41L2H11 growth chamber is designed for growth of Arabidopsis thaliana and low light plant growth applications. SciWhite with red LEDs are dimmable from 10-100% output with up to 405 µmoles/m2/s irradiance @ 6". Ultrasonic humidifier and advanced humidity sensor provide additive humidity. Interior / exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Adjustable air diffuser for temperature uniformity. Two white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.